Learn More
Mouse Nephronectin Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF4298-SP
This item is not returnable.
View return policy
Description
Nephronectin Polyclonal specifically detects Nephronectin in Mouse samples. It is validated for Immunohistochemistry.
Specifications
| Nephronectin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EGFL6L, Nctn, nephronectin, NPNT, POEM | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 255743 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 1-15 ug/mL | |
| AAH68308 | |
| NPNT | |
| Mouse myeloma cell line NS0-derived recombinant mouse Nephronectin Asp20-Cys561 Accession # AAH68308 | |
| 25 μg | |
| Primary | |
| Detects mouse Nephronectin in direct ELISAs. | |
| Mouse | |
| IgG |