missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Nephronectin Antibody, R&D Systems™

Goat Polyclonal Antibody
£170.00 - £516.00
Specifications
| Antigen | Nephronectin |
|---|---|
| Concentration | LYOPH |
| Dilution | Immunohistochemistry 1-15 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18746628
![]() |
R&D Systems
AF4298-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18003472
![]() |
R&D Systems
AF4298 |
100 μg |
£516.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nephronectin Polyclonal specifically detects Nephronectin in Mouse samples. It is validated for Immunohistochemistry.Specifications
| Nephronectin | |
| Immunohistochemistry 1-15 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| EGFL6L, Nctn, nephronectin, NPNT, POEM | |
| NPNT | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| AAH68308 | |
| 255743 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Nephronectin Asp20-Cys561 Accession # AAH68308 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Nephronectin in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title