Learn More
PKN2 Antibody (1D1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-37573-0.025ml
 View more versions of this product
Product Code. 18688885
  • £174.00 / 0.03mL
Description
PKN2 Monoclonal specifically detects PKN2 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PKN2
Monoclonal
Unconjugated
Ascites with 0.03% Sodium Azide
PKN2
Purified recombinant fragment of human PRK2 expressed in E. Coli.
Unpurified
RUO
Primary
Human, Mouse, Rat, Monkey, Primate
Ascites
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
1D1
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin
cardiolipin-activated protein kinase Pak2, EC 2.7.11, EC 2.7.11.13, Pak-2, PAK2, PRK2MGC150606, PRKCL2PRO2042, Protein kinase C-like 2MGC71074, protein kinase N2, Protein-kinase C-related kinase 2, serine/threonine-protein kinase N2
Mouse
140 kDa
0.025 mL
Protein Kinase
5586
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.