missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKN2 Antibody (1D1), Novus Biologicals™
Mouse Monoclonal Antibody
£174.00 - £435.00
Specifications
| Antigen | PKN2 |
|---|---|
| Clone | 1D1 |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18191808
|
Novus Biologicals
NBP2-37573 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18688885
|
Novus Biologicals
NBP2-37573-0.025ml |
0.025 mL |
£174.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PKN2 Monoclonal specifically detects PKN2 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PKN2 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Rat, Monkey, Primate | |
| cardiolipin-activated protein kinase Pak2, EC 2.7.11, EC 2.7.11.13, Pak-2, PAK2, PRK2MGC150606, PRKCL2PRO2042, Protein kinase C-like 2MGC71074, protein kinase N2, Protein-kinase C-related kinase 2, serine/threonine-protein kinase N2 | |
| PKN2 | |
| IgG1 | |
| Unpurified |
| 1D1 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Protein Kinase | |
| Ascites with 0.03% Sodium Azide | |
| 5586 | |
| Purified recombinant fragment of human PRK2 expressed in E. Coli. | |
| Primary | |
| 140 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title