Learn More
E-Cadherin Antibody (CDH1/2208R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-07902-100ug
 View more versions of this product
Product Code. 18670958
  • £439.00 / 100µg
Description
E-Cadherin Monoclonal antibody specifically detects E-Cadherin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
E-Cadherin
Monoclonal
0.2 mg/mL
Western Blot 1-2 μg/ml, Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence 1-2 μg/ml, Immunohistochemistry-Paraffin 1-2 μg/ml, Immunofluorescence 1-2ug/ml
P12830, P12830
Rabbit
Protein A or G purified
RUO
Primary
Human, Mouse
Purified
Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
CDH1/2208R
Unconjugated
10 mM PBS with 0.05% BSA
Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin
Recombinant full-length human E-Cadherin protein (Uniprot: P12830)
100 μg
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction
999
Store at 4°C.