missing translation for 'onlineSavingsMsg'
Learn More

E-Cadherin Antibody (CDH1/2208R), Novus Biologicals™

Rabbit Monoclonal Antibody

£254.00 - £439.00

Specifications

Antigen E-Cadherin
Clone CDH1/2208R
Concentration 0.2 mg/mL
Dilution Western Blot 1-2 μg/ml, Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence 1-2 μg/ml, Immunohistochemistry-Paraffin 1-2 μg/ml, Immunofluorescence 1-2ug/ml
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18670958
View Documents
Novus Biologicals
NBP3-07902-100ug
100 μg
£439.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18644726
View Documents
Novus Biologicals
NBP3-07902-20ug
20 μg
£254.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

E-Cadherin Monoclonal antibody specifically detects E-Cadherin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Specifications

Specifications

E-Cadherin
0.2 mg/mL
Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
Rabbit
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction
10 mM PBS with 0.05% BSA
Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin
Recombinant full-length human E-Cadherin protein (Uniprot: P12830)
Primary
Store at 4°C.
CDH1/2208R
Western Blot 1-2 μg/ml, Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence 1-2 μg/ml, Immunohistochemistry-Paraffin 1-2 μg/ml, Immunofluorescence 1-2ug/ml
Monoclonal
Purified
RUO
Human, Mouse
P12830, P12830
999
IgG
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.