Learn More
PKN2 Rabbit anti-Human, Mouse, Rat, Clone: 5O2G6, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-15891-100UL
This item is not returnable.
View return policy
Description
PKN2 Monoclonal antibody specifically detects PKN2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| PKN2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| 5O2G6 | |
| Western Blot, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| cardiolipin-activated protein kinase Pak2, EC 2.7.11, EC 2.7.11.13, Pak-2, PAK2, PRK2MGC150606, PRKCL2PRO2042, Protein kinase C-like 2MGC71074, protein kinase N2, Protein-kinase C-related kinase 2, serine/threonine-protein kinase N2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PKN2 (Q16513). | |
| 100 μg | |
| Protein Kinase | |
| 5586 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |