missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKN2 Rabbit anti-Human, Mouse, Rat, Clone: 5O2G6, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | PKN2 |
|---|---|
| Clone | 5O2G6 |
| Dilution | Western Blot, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18352725
|
Novus Biologicals
NBP3-15891-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18344322
|
Novus Biologicals
NBP3-15891-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PKN2 Monoclonal antibody specifically detects PKN2 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| PKN2 | |
| Western Blot, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| cardiolipin-activated protein kinase Pak2, EC 2.7.11, EC 2.7.11.13, Pak-2, PAK2, PRK2MGC150606, PRKCL2PRO2042, Protein kinase C-like 2MGC71074, protein kinase N2, Protein-kinase C-related kinase 2, serine/threonine-protein kinase N2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PKN2 (Q16513). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 5O2G6 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5586 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title