Learn More
ASTE1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80291
This item is not returnable.
View return policy
Description
ASTE1 Polyclonal specifically detects ASTE1 in Mouse samples. It is validated for Western Blot.
Specifications
| ASTE1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| asteroid homolog 1 (Drosophila), HT001, MGC129980, protein asteroid homolog 1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 28990 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_079927 | |
| ASTE1 | |
| Synthetic peptide directed towards the middle region of human Aste1. Peptide sequence YHRVLGLLNWLSHFDDPTEALDNVLKSLPKKSRENVKELLCCSMEEYQQS. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Mouse: 100%; Rat: 100%; Guinea pig: 92%; Human: 90%. | |
| Human, Mouse, Rat, Canine, Equine, Guinea Pig | |
| IgG |