Learn More
LYPD4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-58023
This item is not returnable.
View return policy
Description
LYPD4 Polyclonal specifically detects LYPD4 in Human samples. It is validated for Western Blot.
Specifications
| LYPD4 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| LY6/PLAUR domain containing 4, ly6/PLAUR domain-containing protein 4, MGC42718, SMR, sperm membrane receptor | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 147719 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q6UWN0 | |
| LYPD4 | |
| Synthetic peptides corresponding to LYPD4(LY6/PLAUR domain containing 4) The peptide sequence was selected from the N terminal of LYPD4. Peptide sequence MGPQHLRLVQLFCLLGAISTLPRAGALLCYEATASRFRAVAFHNWKWLLM. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Mouse: 78%; Rat: 78%. | |
| Human, Mouse, Rat | |
| IgG |