Learn More
Cadherin-7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59270
This item is not returnable.
View return policy
Description
Cadherin-7 Polyclonal specifically detects Cadherin-7 in Human samples. It is validated for Western Blot.
Specifications
| Cadherin-7 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| cadherin 7, type 2, cadherin-7, CDH7L1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1005 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9ULB5 | |
| CDH7 | |
| Synthetic peptides corresponding to CDH7(cadherin 7, type 2) The peptide sequence was selected from the N terminal of CDH7. Peptide sequence PKFLDGPYTAGVPEMSPVGTSVVQVTATDADDPTYGNSARVVYSILQGQP. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Chicken: 100%; Canine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |