Learn More
DRG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-55158
 View more versions of this product
Product Code. 18214902
  • £381.00 / 100µL
Description
DRG1 Polyclonal specifically detects DRG1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DRG1
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
DRG1
Synthetic peptides corresponding to DRG1(developmentally regulated GTP binding protein 1) The peptide sequence was selected from the middle region of DRG1. Peptide sequence VLKPLGHKKIIENELEGFGIRLNSKPPNIGFKKKDKGGINLTATCPQSEL.
Affinity purified
RUO
4733
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish
IgG
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
developmentally regulated GTP binding protein 1, developmentally regulated GTP-binding protein 1, developmentally-regulated GTP-binding protein 1, DKFZp434N1827, DRG-1, NEDD3, NEDD-3, Neural precursor cell expressed developmentally down-regulated protein 3, neural precursor cell expressed, developmentally down-regulated 3
Rabbit
26 kDa
100 μL
Primary
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.