Learn More
SEC6 Antibody (9H5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Brand:  Novus Biologicals NBP1-97500-0.025MG
 View more versions of this product
Product Code. 18006637
  • £145.00 / 25µg
Description
SEC6 Monoclonal specifically detects SEC6 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Hamster, Monkey, Rabbit, Sheep, Yeast samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SEC6
Monoclonal
Unconjugated
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
EXOC3
Recombinant rat Sec6.
0.025 mg
Primary
Chicken, Mouse, Sheep, Rabbit, Pig, Monkey, Rat, Human, Canine, Bovine, Hamster
Purified
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry, Immunocytochemistry
9H5
Western Blot 1:250, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunoblotting
exocyst complex component 3, Exocyst complex component Sec6, Sec 6 homolog, SEC6, SEC6L1, SEC6-like 1, SEC6-like 1 (S. cerevisiae), Sec6p
Mouse
Protein G purified
RUO
11336
Store at -20C. Avoid freeze-thaw cycles.
IgG2a