missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC6 Antibody (9H5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
£145.00 - £564.00
Specifications
| Antigen | SEC6 |
|---|---|
| Clone | 9H5 |
| Dilution | Western Blot 1:250, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunoblotting |
| Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18006637
|
Novus Biologicals
NBP1-97500-0.025MG |
0.025 mg |
£145.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18058828
|
Novus Biologicals
NBP1-97500-0.05MG |
0.05 mg |
£281.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18105700
|
Novus Biologicals
NBP1-97500-0.2MG |
0.2 mg |
£564.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SEC6 Monoclonal specifically detects SEC6 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Hamster, Monkey, Rabbit, Sheep, Yeast samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
| SEC6 | |
| Western Blot 1:250, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunoblotting | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| 11336 | |
| Recombinant rat Sec6. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 9H5 | |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Chicken, Mouse, Sheep, Rabbit, Pig, Monkey, Rat, Human, Canine, Bovine, Hamster | |
| exocyst complex component 3, Exocyst complex component Sec6, Sec 6 homolog, SEC6, SEC6L1, SEC6-like 1, SEC6-like 1 (S. cerevisiae), Sec6p | |
| EXOC3 | |
| IgG2a | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title