Learn More
STEMCELL Technologies RoboSep™ Human Naïve CD4+ T Cell Isolation Kit II
Description
- The EasySep™ Human Naïve CD4+ T Cell Isolation Kit II is designed to isolate naïve CD4+ T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection
- The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
- The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
- This kit replaces the EasySep™ Human Naïve CD4+ T Cell Enrichment Kit (Catalog #19155) and EasySep™ Human Naïve CD4+ T Cell Isolation Kit (Catalog #19555) for even faster cell isolations
Specifications
Specifications
| Cell Type | T Cells, T Cells, CD4+ |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |