Learn More
Agilent BioTek 96-tube Dual-action Tube-in-tube Aspiration Manifold
Product Code. 16592062 Shop All Agilent BioTek Products
Product Code. 16592062
Brand: Agilent BioTek 1172046S
96-tube Dual-action Tube-in-tube Aspiration Manifold is for 384-well washing with buffers not containing surfactant