Learn More
MedChemExpress NBD-F
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
NBD-F reacts with primary or secondary amines to produce a fluorescent product and used for analysis of amino acids and low molecular weight amines.
Specifications
Specifications
| Quantity | 100 mg |
| Product Type | Dye Reagents |
| Molecular Weight (g/mol) | 183.1 g/mol |
| Solubility | DMSO : 100 mg/mL (546.15 mM; Need ultrasonic) |
| Content And Storage | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Shipping Condition | Room temperature |
| Target | HBV |
| CAS | 29270-56-2 |
| Synonym | 4-Fluoro-7-nitrobenzofurazan |