Learn More
D-Methionine methyl ester hydrochloride, 98%
Codice prodotto. 15420109
Click to view available options
Quantity:
5 g
25 g
100 g
Dimensione della confezione:
100g
25g
5g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15420109
Marca: Thermo Scientific Alfa Aesar H63422.14
Request Bulk or Custom Formats
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
CAS: 69630-60-0 | C6H14ClNO2S | 199.69 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69630-60-0
Molecular Formula C6H14ClNO2S
Molecular Weight (g/mol) 199.69
MDL Number MFCD00070385
InChI Key MEVUPUNLVKELNV-NUBCRITNSA-N
Synonym h-d-met-ome.hcl, d-methionine methyl ester hydrochloride, h-d-met-ome hcl, methyl d-methioninate-hydrogen chloride 1/1, methyl 2r-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, h-d-met-omecl, d-methionine methyl ester hcl, methyl d-methioninate hydrochloride, d methionine methyl ester hydrochloride
PubChem CID 12888616
IUPAC Name methyl (2R)-2-amino-4-methylsulfanylbutanoate;hydrochloride
SMILES Cl.COC(=O)[C@H](N)CCSC
Specifications
Quantity 25 g
Formula Weight 199.7
Percent Purity 98%
Physical Form Solid
Chemical Name or Material D-Methionine methyl ester hydrochloride
RUO – Research Use Only