accessibility menu, dialog, popup

UserName

CAS RN 69630-60-0

CH3OO(R)SH3CNH2HCl

CAS RN 69630-60-0

IUPAC Name: methyl 2-amino-4-(methylsulfanyl)butanoate hydrochloride
Synonyms: methyl 2-amino-4-(methylsulfanyl)butanoate hydrochloride
Molecular Weight (g/mol): 199.69
Molecular Formula: C6H14ClNO2S
InChi Key: MEVUPUNLVKELNV-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(N)CCSC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)