Learn More
Thermo Scientific™ Oxoid™ Violet Red Bile Agar (VRB Agar)
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 20
Description
Enhance the detection of enteric bacteria with Violet Red Bile Agar (VRB Agar), providing selective isolation for food and water testing.
Specifications
Specifications
| Color | Violet |
| Content And Storage | 2°C to 12°C |
| Format | 90 mm Plate |
| Manufacturing Origin | Germany |
| Product Type | Agar Plate |
| Fill Weight | 17 g |
| Quantity | 10 Plates |