Learn More
Fisherbrand™ accuTupH Plus™ Rugged Bulb Variable Temperature pH Combination Electrode
Shop All Fisherbrand Products
Click to view available options
Connector Types:
BNC
Unit Size:
Each
Description
Can be optimal for lab use and precise Q.C. measurements.
- 0 to 14 ph range
- -5 to +100°C range
- 102mmL, 10mm dia.
Specifications
Specifications
| Connector Types | BNC |
| Junction | Double |
| Material | Glass |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | -5°C to +100°C |
| Type | Combination pH Electrode |
| Filling Solution Type | Liquid |
| For Use With (Application) | Wide Temperature Variation |
| Temperature (English) Operating | 23°F to 212°F |
| Temperature (Metric) Range | -5°C to +100°C |
| Show More |