Learn More
VICI Jour Chromatography™ Polypropylene Flanged Fingertight Nuts, 1/8 in.
For flanged connections
Brand:  VICI Jour Chromatography™ JR-201580WH-10
Code : 15
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12995021
  • £59.20 / Pack of 10
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • Compatible to all 1/4"-28 flat bottom ports
  • For 1/8" OD tubing
  • Fiber glass reinforced PP with PP washers
Specifications
Specifications
Nut
10 Pack
Polypropylene
White
For 1.41 mm Tubing