Learn More
VICI Jour Chromatography™ Polypropylene Flanged Fingertight Nuts, 1/8 in.
For flanged connections
£59.20
Specifications
Type Nut
Packaging Quantity 10 Pack
For Use With For 1.41 mm Tubing
Material Polypropylene
View More Specs
Products 2
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
12995021
View Documents Promotion Details
VICI Jour Chromatography™
JR-201580WH-10
White
£59.20
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Add to basket
12905031
View Documents Promotion Details
VICI Jour Chromatography™
JR-201580BK-10
Black
£59.20
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Add to basket
Description
Description
  • Compatible to all 1/4"-28 flat bottom ports
  • For 1/8" OD tubing
  • Fiber glass reinforced PP with PP washers
Specifications
Specifications
Nut
10 Pack
For 1.41 mm Tubing
Polypropylene