Learn More
VICI Jour Chromatography™ Polypropylene Flanged Fingertight Nuts, 1/8 in.
For flanged connections
Brand: VICI Jour Chromatography™ JR-201580BK-10
Code : 15
Additional Details : Weight : 1.00000kg
Description
- Compatible to all 1/4"-28 flat bottom ports
- For 1/8" OD tubing
- Fiber glass reinforced PP with PP washers
Specifications
Nut | |
10 Pack | |
Polypropylene |
Black | |
For 1.41 mm Tubing |