missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VICI Valco Instruments™ Stainless Steel Mobile Phase Filter
Brand: VICI Valco Instruments™ JR-4676-2
Code : 15
Additional Details : Weight : 1.00000kg
Specifications
2 μm | |
Mobile Phase Filter | |
19 mm | |
Stainless Steel |