Learn More
VICI Valco Instruments™ Stainless Steel Mobile Phase Filter
£54.06 - £59.25
Product Code | Brand | Pore Size | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
12915051
|
VICI Valco Instruments™
JR-4676-10 |
10 μm | 19 mm |
£54.06
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
||||
12905051
|
VICI Valco Instruments™
JR-4676-2 |
2 μm | 19 mm |
£55.65
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
||||
12737393
|
VICI Valco Instruments™
JR-4677-10 |
10 μm | 22 mm |
£59.20
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
||||
12925051
|
VICI Valco Instruments™
JR-4677-2 |
2 μm | 22 mm |
£59.25
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
||||
Specifications
Mobile Phase Filter | |
Stainless Steel |