missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech Viability/Cytotoxicity Assay Kit for Animal Live & Dead Cells (Calcein AM, PI Method)
Shop All Proteintech Products
Click to view available options
Quantity:
150 Tests
300 Tests
Unit Size:
150 tests
300 tests
Description
- The two probes in the kit effect cell viability by measuring intracellular esterase activity and plasma membrane integrity.
- This kit can be used for fluorescence microscopes, flow cytometers, microplate readers and other fluorescence detection systems.
- This kit can be used for most eukaryotic mammalian samples, but not to fungi and yeast.
Specifications
Specifications
| For Use With (Application) | Flow Cytometry |
| Emission | 617/517 |
| Quantity | 300 Tests |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction