missing translation for 'onlineSavingsMsg'
Learn More

Proteintech Viability/Cytotoxicity Assay Kit for Animal Live & Dead Cells (Calcein AM, EthD-1 Method)

Product Code. 17581176 Shop All Proteintech Products
Click to view available options
Quantity:
150 Tests
300 Tests
Unit Size:
150 tests
300 tests
This item is not returnable. View return policy

Product Code. 17581176

Brand: Proteintech PF00008300T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Viability/Cytotoxicity Assay Kit for Animal Live Dead Cells (Calcein AM, EthD-I method) provides dual fluorescence staining for the detection of living and dead cells.

  • The two probes in the kit effect cell viability by measuring intracellular esterase activity and plasma membrane integrity.
  • This kit can be used for fluorescence microscopes, flow cytometers, microplate readers and other fluorescence detection systems.
  • This kit can be used for most eukaryotic mammalian samples, but not to fungi and yeast.

Specifications

For Use With (Application) Flow Cytometry
Emission 617/517
Quantity 300 Tests
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.