missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress VH032-PEG5-C6-Cl
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Description
The connector of linker is Halogen group. VH032-PEG5-C6-Cl incorporates the VH032 based VHL ligand and 5-unit PEG linker. VH032-PEG5-C6-Cl is capable of inducing the degradation of GFP-HaloTag7 in cell-based assays.
Specifications
Specifications
| Molecular Weight (g/mol) | 783.41 g/mol |
| CAS | 1799506-06-1 |
| Content And Storage | -80°C, protect from light, stored under nitrogen |
| Shipping Condition | Shipping with dry ice |
| Quantity | 25 mg |
| Synonym | HaloPROTAC 2 |
| Solubility | DMSO : 270 mg/mL (344.65 mM; Need ultrasonic) ∣ Ethanol : 100 mg/mL (127.65 mM; Need ultrasonic) |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction