missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress VH032-PEG5-C6-Cl
Description
The connector of linker is Halogen group. VH032-PEG5-C6-Cl incorporates the VH032 based VHL ligand and 5-unit PEG linker. VH032-PEG5-C6-Cl is capable of inducing the degradation of GFP-HaloTag7 in cell-based assays.
Specifications
Specifications
| Molecular Weight (g/mol) | 783.41 g/mol |
| CAS | 1799506-06-1 |
| Content And Storage | -80°C, protect from light, stored under nitrogen |
| Shipping Condition | Shipping with dry ice |
| Quantity | 10 mg |
| Synonym | HaloPROTAC 2 |
| Solubility | DMSO : 270 mg/mL (344.65 mM; Need ultrasonic) ∣ Ethanol : 100 mg/mL (127.65 mM; Need ultrasonic) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?