missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGLUT2 Antibody (S29-29), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59330
This item is not returnable.
View return policy
Description
VGLUT2 Monoclonal specifically detects VGLUT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| VGLUT2 | |
| Monoclonal | |
| Unconjugated | |
| PBS pH7.4, 50% glycerol with 0.09% Sodium Azide | |
| SLC17A6 | |
| Fusion protein amino acids 501-582 (cytoplasmic C-terminus) of rat VGlut2 | |
| Protein G purified | |
| RUO | |
| 57084 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S29-29 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| differentiation-associated BNPI, differentiation-associated Na(+)-dependent inorganic phosphate cotransporter, differentiation-associated Na-dependent inorganic phosphate cotransporter, DNPI, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6, solute carrier family 17 member 6, vesicular glutamate transporter 2, VGLUT2 | |
| Mouse | |
| 60 kDa | |
| 100 μg | |
| Primary | |
| Detects 60kDa. | |
| Store at -20C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction