missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGLUT2 Antibody (S29-29), Novus Biologicals™
Mouse Monoclonal Antibody
£181.00 - £474.00
Specifications
| Antigen | VGLUT2 |
|---|---|
| Clone | S29-29 |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18242462
|
Novus Biologicals
NBP2-59330 |
100 μg |
£474.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18622497
|
Novus Biologicals
NBP2-59330-25ug |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VGLUT2 Monoclonal specifically detects VGLUT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| VGLUT2 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| 57084 | |
| Fusion protein amino acids 501-582 (cytoplasmic C-terminus) of rat VGlut2 | |
| Primary | |
| Detects 60kDa. |
| S29-29 | |
| Monoclonal | |
| Purified | |
| RUO | |
| differentiation-associated BNPI, differentiation-associated Na(+)-dependent inorganic phosphate cotransporter, differentiation-associated Na-dependent inorganic phosphate cotransporter, DNPI, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6, solute carrier family 17 member 6, vesicular glutamate transporter 2, VGLUT2 | |
| SLC17A6 | |
| IgG1 | |
| Protein G purified | |
| 60 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title