missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGLL4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£186.00 - £375.00
Specifications
| Antigen | VGLL4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637261
|
Novus Biologicals
NBP2-93539-0.02ml |
0.02 mL |
£186.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640771
|
Novus Biologicals
NBP2-93539-0.1ml |
0.1 mL |
£375.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VGLL4 Polyclonal antibody specifically detects VGLL4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| VGLL4 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| KIAA0121transcription cofactor vestigial-like protein 4, vestigial like 4 (Drosophila), Vestigial-like 4, Vgl-4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-287 of mouse Vgll4 (NP_808351.1). MLFMKMDLLNYQYLDKMNNNIGVLCYEGEASLRGEPRMQTLPVASALSSHRTGPPPISPSKRKFSMEPGDKDLDCENDHVSKMSRIFSPHLNKTVNGDCRRDPRERSRSPIERAAAPAVSLHGGHLYASLPSLMEQPLALTKNSSDTGRSAVERQQNRPSVITCASAGARNCNLSHCPIAHSGCSAPGSASYRRPPSATATCDPVVEEHFRRSLGKNYKEPEPAPNSVSITGSVDDHFAKALGDTWLQIKAAKDSASSSPESASRRGQPASPTAHMVSHSHSPSVVS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 9686 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title