missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGLL4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93539-0.02ml
This item is not returnable.
View return policy
Description
VGLL4 Polyclonal antibody specifically detects VGLL4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| VGLL4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| KIAA0121transcription cofactor vestigial-like protein 4, vestigial like 4 (Drosophila), Vestigial-like 4, Vgl-4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-287 of mouse Vgll4 (NP_808351.1). MLFMKMDLLNYQYLDKMNNNIGVLCYEGEASLRGEPRMQTLPVASALSSHRTGPPPISPSKRKFSMEPGDKDLDCENDHVSKMSRIFSPHLNKTVNGDCRRDPRERSRSPIERAAAPAVSLHGGHLYASLPSLMEQPLALTKNSSDTGRSAVERQQNRPSVITCASAGARNCNLSHCPIAHSGCSAPGSASYRRPPSATATCDPVVEEHFRRSLGKNYKEPEPAPNSVSITGSVDDHFAKALGDTWLQIKAAKDSASSSPESASRRGQPASPTAHMVSHSHSPSVVS | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9686 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction