missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGCNL1 Antibody (S187-7), Novus Biologicals™
Mouse Monoclonal Antibody
£176.00
Specifications
| Antigen | VGCNL1 |
|---|---|
| Clone | S187-7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1μg/mL, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Description
VGCNL1 Monoclonal specifically detects VGCNL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| VGCNL1 | |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| bA430M15.1, CanIonFLJ44659, FLJ23913, FLJ44764, four repeat voltage-gated ion channel, MGC74524, sodium leak channel non-selective protein, sodium leak channel, non-selective, VGCNL1, voltage gated channel like 1, Voltage gated channel-like protein 1 | |
| NALCN | |
| IgG1 | |
| Protein G purified | |
| Detects approx 200kDa. |
| S187-7 | |
| Western Blot 1μg/mL, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q6Q760 | |
| 259232 | |
| Fusion protein amino acids 1659-1738 (cytoplasmic C-terminus) of rat NALCN | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title