missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGCNL1 Antibody (S187-7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22410-0.025mg
This item is not returnable.
View return policy
Description
VGCNL1 Monoclonal specifically detects VGCNL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| VGCNL1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1μg/mL, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| bA430M15.1, CanIonFLJ44659, FLJ23913, FLJ44764, four repeat voltage-gated ion channel, MGC74524, sodium leak channel non-selective protein, sodium leak channel, non-selective, VGCNL1, voltage gated channel like 1, Voltage gated channel-like protein 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 259232 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| S187-7 | |
| Unconjugated | |
| Q6Q760 | |
| NALCN | |
| Fusion protein amino acids 1659-1738 (cytoplasmic C-terminus) of rat NALCN | |
| 0.025 mg | |
| Primary | |
| Detects approx 200kDa. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction