missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VG5Q Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£171.00 - £362.00
Specifications
| Antigen | VG5Q |
|---|---|
| Dilution | Western Blot 1:500, ELISA reported in scientific literature, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18066094
|
Novus Biologicals
NB100-454SS |
0.025 mL |
£171.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18096164
|
Novus Biologicals
NB100-454 |
0.1 mL |
£362.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VG5Q Polyclonal specifically detects VG5Q in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| VG5Q | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Hypoxia | |
| Whole antisera with No Preservative | |
| Angiogenic factor VG5Q, angiogenic factor with G patch and FHA domains 1, FLJ10283hVG5Q, GPATC7G patch domain-containing protein 7, GPATCH7HUS84971, HSU84971, VG5QVasculogenesis gene on 5q protein | |
| AGGF1 | |
| IgG | |
| Unpurified | |
| This is specific for human VG5Q. |
| Western Blot 1:500, ELISA reported in scientific literature, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| Q8N302 | |
| 55109 | |
| A synthetic peptide mapping to an epitope within residues 500-600 of the human VG5Q protein. [UniProt# Q8N302] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 100 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title