missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VG5Q Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-454
This item is not returnable.
View return policy
Description
VG5Q Polyclonal specifically detects VG5Q in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| VG5Q | |
| Polyclonal | |
| Western Blot 1:500, ELISA reported in scientific literature, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin | |
| Q8N302 | |
| AGGF1 | |
| A synthetic peptide mapping to an epitope within residues 500-600 of the human VG5Q protein. [UniProt# Q8N302] | |
| Unpurified | |
| RUO | |
| Primary | |
| This is specific for human VG5Q. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Whole antisera with No Preservative | |
| Angiogenic factor VG5Q, angiogenic factor with G patch and FHA domains 1, FLJ10283hVG5Q, GPATC7G patch domain-containing protein 7, GPATCH7HUS84971, HSU84971, VG5QVasculogenesis gene on 5q protein | |
| Rabbit | |
| 100 kDa | |
| 0.1 mL | |
| Angiogenesis, Hypoxia | |
| 55109 | |
| Human | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction