missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain Versilic™ Silicone Stoppers, 40mmH

Brand:  Saint-Gobain 26305402

Code : A0

Additional Details : Weight : 0.15300kg

 View more versions of this product

Product Code. 11774578

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Biologically neutral and non-toxic silicone caps for closing tubes and flasks
  • Odorless, tasteless, non-adherent and resistant to aging
  • Nontoxic, biologically neutral
  • FDA/BGA approved
  • For test tubes
  • Easy to install
Specifications

Specifications

Silicone Versilic™ Cap
47 mm
2/Pk.
–60 to 200°C
FDA compliant food contact / BFR
40mm
55 mm
Silicone
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers