missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFR1/Flt-1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£118.00
Specifications
| Antigen | VEGFR1/Flt-1 |
|---|---|
| Concentration | LYOPH |
| Applications | Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
VEGFR1/Flt-1 Polyclonal specifically detects VEGFR1/Flt-1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| VEGFR1/Flt-1 | |
| Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot, ELISA | |
| Unconjugated | |
| Human | |
| 2321 | |
| IgG | |
| soluble VEGFR-1/Flt-1 |
| LYOPH | |
| Polyclonal | |
| Rabbit | |
| EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1 | |
| FLT1 | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title