missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFR1/Flt-1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-18631-0.02MG
This item is not returnable.
View return policy
Description
VEGFR1/Flt-1 Polyclonal specifically detects VEGFR1/Flt-1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| VEGFR1/Flt-1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| FLT1 | |
| Peptide: GEHCNKKAVFSRISKFKSTR NDSTTQSNVKH. | |
| 0.02 mg | |
| Angiogenesis, Cancer, HIF Target Genes, Hypoxia, Stem Cell Markers | |
| 2321 | |
| Reconstitute in sterile water to a concentration of 0.1-1.0mg/mL. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot, ELISA | |
| LYOPH | |
| Western Blot 1-5 ug, ELISA 2.5 ug/ml, Immunocytochemistry/Immunofluorescence 1-5 ug, Immunoprecipitation 1-5 ug/ml | |
| EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| soluble VEGFR-1/Flt-1 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction