missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF-B Antibody (RM0008-6E72) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£168.00 - £444.00
Specifications
| Antigen | VEGF-B |
|---|---|
| Clone | RM0008-6E72 |
| Concentration | LYOPH |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18253741
|
Novus Biologicals
NB110-61024-0.025MG |
0.025 mg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18107269
|
Novus Biologicals
NB110-61024 |
0.1 mg |
£444.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VEGF-B Monoclonal specifically detects VEGF-B in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| VEGF-B | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| 7423 | |
| IgG2 | |
| Protein G purified |
| RM0008-6E72 | |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Angiogenesis, Cell Cycle and Replication | |
| vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRFVEGFL | |
| VEGFB | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title