missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF-B Antibody (RM0008-6E72) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB110-61024-0.025MG
This item is not returnable.
View return policy
Description
VEGF-B Monoclonal specifically detects VEGF-B in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| VEGF-B | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:400-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRFVEGFL | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| VEGFB (RM0008-6E72) | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| RM0008-6E72 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| VEGFB | |
| This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with purified mouse VEGF-B (186). | |
| 0.025 mg | |
| Angiogenesis, Cell Cycle and Replication | |
| 7423 | |
| Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction