missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF Antibody (9P0F4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-33160-20ul
This item is not returnable.
View return policy
Description
VEGF Monoclonal antibody specifically detects VEGF in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| VEGF | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH7.3), 50% glycerol | |
| Mouse | |
| Affinity purified | |
| RUO | |
| 7422 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| ELISA, Western Blot | |
| 9P0F4 | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 207-327 of human VEGF (P15692).,, Sequence: APMAEGGGQNHHEVVKFMDVYQRSYCHPIETLVDIFQEYPDEIEYIFKPSCVPLMRCGGCCNDEGLECVPTEESNITMQIMRIKPHQGQHIGEMSFLQHNKCECRPKKDRARQENCDKPRR | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto