missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF Antibody (9P0F4), Novus Biologicals™
Mouse Monoclonal Antibody
£145.00 - £351.00
Specifications
| Antigen | VEGF |
|---|---|
| Clone | 9P0F4 |
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30141288
|
Novus Biologicals (Bio-Techne)
NBP3-33160-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30145008
|
Novus Biologicals (Bio-Techne)
NBP3-33160 |
100 μL |
£351.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VEGF Monoclonal antibody specifically detects VEGF in Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| VEGF | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH7.3), 50% glycerol | |
| 7422 | |
| IgG1 κ | |
| Affinity purified |
| 9P0F4 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse, Rat | |
| MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 207-327 of human VEGF (P15692).,, Sequence: APMAEGGGQNHHEVVKFMDVYQRSYCHPIETLVDIFQEYPDEIEYIFKPSCVPLMRCGGCCNDEGLECVPTEESNITMQIMRIKPHQGQHIGEMSFLQHNKCECRPKKDRARQENCDKPRR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title