missing translation for 'onlineSavingsMsg'
Learn More

VEGF Antibody (9P0F4), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP3-33160

 View more versions of this product

Product Code. 30145008

  • £351.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

VEGF Monoclonal antibody specifically detects VEGF in Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VEGF
Monoclonal
Unconjugated
PBS (pH7.3), 50% glycerol
Mouse
Affinity purified
RUO
7422
Store at -20°C. Avoid freeze-thaw cycles.
IgG1 κ
ELISA, Western Blot
9P0F4
Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
Recombinant fusion protein containing a sequence corresponding to amino acids 207-327 of human VEGF (P15692).,, Sequence: APMAEGGGQNHHEVVKFMDVYQRSYCHPIETLVDIFQEYPDEIEYIFKPSCVPLMRCGGCCNDEGLECVPTEESNITMQIMRIKPHQGQHIGEMSFLQHNKCECRPKKDRARQENCDKPRR
100 μL
Primary
Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.