missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF 165 Rabbit anti-Human, Clone: R012, Novus Biologicals™
Rabbit Monoclonal Antibody
£393.00 - £522.00
Specifications
| Antigen | VEGF 165 |
|---|---|
| Clone | R012 |
| Applications | Neutralization |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18219087
|
Novus Biologicals
NBP2-89779-200UG |
200 μg |
£393.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18237737
|
Novus Biologicals
NBP2-89779-500UG |
500 μg |
£522.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VEGF 165 Monoclonal specifically detects VEGF 165 in Human samples. It is validated for Neutralization.Specifications
| VEGF 165 | |
| Neutralization | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Biologically Active Proteins | |
| 0.2 um filtered solution in PBS | |
| 7422 | |
| IgG | |
| Protein A purified |
| R012 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
| Recombinant Human VEGF 165 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title