missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF 165 Rabbit anti-Human, Clone: R012, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89779-500UG
This item is not returnable.
View return policy
Description
VEGF 165 Monoclonal specifically detects VEGF 165 in Human samples. It is validated for Neutralization.
Specifications
| VEGF 165 | |
| Monoclonal | |
| Unconjugated | |
| MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
| Recombinant Human VEGF 165 protein | |
| 500 μg | |
| Angiogenesis, Biologically Active Proteins | |
| 7422 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Neutralization | |
| R012 | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction