missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VDR/NR1I1/Vitamin D Receptor Antibody (3C7), Novus Biologicals™
Rabbit Monoclonal Antibody
£269.00 - £413.00
Specifications
| Antigen | VDR/NR1I1/Vitamin D Receptor |
|---|---|
| Clone | 3C7 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30027492
|
Novus Biologicals
NBP3-26255-50ul |
50 μL |
£269.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30039164
|
Novus Biologicals
NBP3-26255-100ul |
100 μL |
£413.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VDR/NR1I1/Vitamin D Receptor Monoclonal antibody specifically detects VDR/NR1I1/Vitamin D Receptor in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| VDR/NR1I1/Vitamin D Receptor | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| NR1I1Nuclear receptor subfamily 1 group I member 11,25-dihydroxyvitamin D3 receptor, vitamin D1,25- dihydroxyvitamin D3 receptor, vitamin D3 receptor | |
| A synthesized peptide derived from Human VDR/NR1I1/Vitamin D Receptor [UniProt P11473] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 3C7 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 7421 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title