missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VDR/NR1I1/Vitamin D Receptor Antibody (3C7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26255-50ul
This item is not returnable.
View return policy
Description
VDR/NR1I1/Vitamin D Receptor Monoclonal antibody specifically detects VDR/NR1I1/Vitamin D Receptor in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| VDR/NR1I1/Vitamin D Receptor | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| 3C7 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| NR1I1Nuclear receptor subfamily 1 group I member 11,25-dihydroxyvitamin D3 receptor, vitamin D1,25- dihydroxyvitamin D3 receptor, vitamin D3 receptor | |
| A synthesized peptide derived from Human VDR/NR1I1/Vitamin D Receptor [UniProt P11473] | |
| 50 μL | |
| Signal Transduction | |
| 7421 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction